missing translation for 'onlineSavingsMsg'
Learn More

3-Aminooxetane-3-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog Number 15433798
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy
138650-24-5
C4H7NO3
117.104
MFCD11035888
LXRNYRAHXDUNKR-UHFFFAOYSA-N
3-oxetanecarboxylic acid, 3-amino, 3-amino-oxetane-3-carboxylic acid, acmc-20a7wi, 3-amino-3-oxetanecarboxylic acid, 3-aminooxetane-3-carboxylic acid
15024256
3-aminooxetane-3-carboxylic acid
C1C(CO1)(C(=O)O)N
This item is not returnable. View return policy
138650-24-5
C4H7NO3
117.104
MFCD11035888
LXRNYRAHXDUNKR-UHFFFAOYSA-N
3-oxetanecarboxylic acid, 3-amino, 3-amino-oxetane-3-carboxylic acid, acmc-20a7wi, 3-amino-3-oxetanecarboxylic acid, 3-aminooxetane-3-carboxylic acid
15024256
3-aminooxetane-3-carboxylic acid
C1C(CO1)(C(=O)O)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 205°C to 210°C
Color White
Quantity 250 mg
Formula Weight 117.1
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 3-Aminooxetane-3-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'