missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(Aminomethyl)benzoic acid hydrochloride, 95%

Brand:  Alfa Aesar™ H64546.MD

Code : NEW

Additional Details : CAS Number : 876-03-9 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15418976

  • £9.90 / 250mg


Estimated Shipment date
from Supplier 05-07-2018
Add to basket

Description

Description

Applications
It is used as pharmaceutical intermediate. Derivatives of benzylamine and benzamidine were found to be competitive inhibitors of the proteolytic enzymes trypsin, plasmin, and thrombin.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

3-(Aminomethyl)benzoic acid hydrochloride
187.62
3697656
876-03-9
MFCD03791117

Specifications

3-(Aminomethyl)benzoic acid hydrochloride
242° to 244°C
95%
187.62
3697656
876-03-9
Solid
250mg
MFCD03791117
Videos
SDS