missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Aminomethyl)benzeneboronic acid hydrochloride, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
3-(Aminomethyl)benzeneboronic acid hydrochloride is used as intermediates. Hydroxybenzene boronic acids are involved in suzuki miyaura reactions. It is a arylboronic acid. 3-Fluoro-4-hydroxybenzeneboronic acid, can be an effective catalyst for amidation and esterification of carboxylic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(Aminomethyl)benzeneboronic acid hydrochloride is used as intermediates. Hydroxybenzene boronic acids are involved in suzuki miyaura reactions. It is a arylboronic acid. 3-Fluoro-4-hydroxybenzeneboronic acid, can be an effective catalyst for amidation and esterification of carboxylic acids.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerate.
3-(Aminomethyl)benzeneboronic acid hydrochloride is used as intermediates. Hydroxybenzene boronic acids are involved in suzuki miyaura reactions. It is a arylboronic acid. 3-Fluoro-4-hydroxybenzeneboronic acid, can be an effective catalyst for amidation and esterification of carboxylic acids.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerate.
Specifications
Melting Point | 248°C to 250°C |
Quantity | 250 mg |
Solubility Information | Slightly soluble in water. |
Formula Weight | 187.43 |
Percent Purity | 96% |
Chemical Name or Material | 3-(Aminomethyl)benzeneboronic acid hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'