missing translation for 'onlineSavingsMsg'
Learn More

3-Aminocarbonylphenylboronic acid, 97%, ACROS Organics™

Brand:  Acros Organics 444890050

Code : 29

Additional Details : CAS Number : 351422-73-6 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 11916091

  • £125.90 / 5g


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

3-Aminocarbonylphenylboronic acid
97%
164.96
97%
C7H8BNO3

Specifications

3-Aminocarbonylphenylboronic acid
351422-73-6
232.0° to 238.0°C
5g
97%
C7H8BNO3
Glass bottle
96% min. consistent with structure (1H-NMR)
White to off-white
Solid
97%
164.96
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Aminocarbonylphenylboronic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS