missing translation for 'onlineSavingsMsg'
Learn More

3-Aminocarbonylphenylboronic acid, 97%, Thermo Scientific™

Catalog Number 11906091
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
351422-73-6
C7H8BNO3
164.96
MFCD03411948
WDGWHKRJEBENCE-UHFFFAOYSA-N
3-aminocarbonylphenylboronic acid, 3-carbamoylphenyl boronic acid, benzamide-3-boronic acid, 3-carbamoylbenzeneboronic acid, 3-aminocarbonyphenylboronic acid, 3-aminocarbonyl benzeneboronic acid, 3-aminocarbonylphenyl boronic acid, 3-carboxamidephenylboronic acid, 3-aminocarbonyl phenylboronic acid, 3-aminocarbonyl phenyl boronic acid
2737814
(3-carbamoylphenyl)boronic acid
NC(=O)C1=CC=CC(=C1)B(O)O
This item is not returnable. View return policy
351422-73-6
C7H8BNO3
164.96
MFCD03411948
WDGWHKRJEBENCE-UHFFFAOYSA-N
3-aminocarbonylphenylboronic acid, 3-carbamoylphenyl boronic acid, benzamide-3-boronic acid, 3-carbamoylbenzeneboronic acid, 3-aminocarbonyphenylboronic acid, 3-aminocarbonyl benzeneboronic acid, 3-aminocarbonylphenyl boronic acid, 3-carboxamidephenylboronic acid, 3-aminocarbonyl phenylboronic acid, 3-aminocarbonyl phenyl boronic acid
2737814
(3-carbamoylphenyl)boronic acid
NC(=O)C1=CC=CC(=C1)B(O)O
Melting Point 232.0°C to 238.0°C
Color White
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 164.96
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Aminocarbonylphenylboronic acid
compliance-icons
Product Identifier
  • 3-Aminocarbonylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'