missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-3586352
27.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4403-70-7
C7H10N2
122.17
MFCD00078355
ZDBWYUOUYNQZBM-UHFFFAOYSA-N
3-aminobenzylamine, 3-aminomethyl aniline, m-aminobenzylamine, benzenemethanamine, 3-amino, 3-aminomethyl-aniline, pubchem5802, alpha-amino-m-toluidine, 3-aminomethyl phenylamine, 3-aminophenyl methylamine
4628831
3-(aminomethyl)aniline
NCC1=CC(N)=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4403-70-7
C7H10N2
122.17
MFCD00078355
ZDBWYUOUYNQZBM-UHFFFAOYSA-N
3-aminobenzylamine, 3-aminomethyl aniline, m-aminobenzylamine, benzenemethanamine, 3-amino, 3-aminomethyl-aniline, pubchem5802, alpha-amino-m-toluidine, 3-aminomethyl phenylamine, 3-aminophenyl methylamine
4628831
3-(aminomethyl)aniline
NCC1=CC(N)=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 36°C to 39°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula NH2CH2C6H4NH2
Quantity 5 g
Formula Weight 122.17
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 3-Aminobenzylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Aminobenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'