missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminobenzyl alcohol, 98+%, Thermo Scientific Chemicals
69.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
3-Aminobenzyl alcohol, is used as a reagent in the synthesis of quinone analogs as dynamin GTPase inhibitors. Also used as a reagent in the synthesis of pyrrolo[2,1-f][1,2,4]triazines as novel hedgehog signaling pathway inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Aminobenzyl alcohol, is used as a reagent in the synthesis of quinone analogs as dynamin GTPase inhibitors. Also used as a reagent in the synthesis of pyrrolo[2,1-f][1,2,4]triazines as novel hedgehog signaling pathway inhibitors.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
3-Aminobenzyl alcohol, is used as a reagent in the synthesis of quinone analogs as dynamin GTPase inhibitors. Also used as a reagent in the synthesis of pyrrolo[2,1-f][1,2,4]triazines as novel hedgehog signaling pathway inhibitors.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 91°C to 95°C |
Quantity | 25 g |
Beilstein | 2205844 |
Sensitivity | Air and light sensitive |
Solubility Information | Soluble in water. |
Formula Weight | 123.16 |
Percent Purity | ≥98% |
Chemical Name or Material | 3-Aminobenzyl alcohol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'