missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzotrifluoride 98%, ACROS Organics™

Brand:  Acros Organics 103191000

Code : 29

Additional Details : CAS Number : 98-16-8 Weight : 0.18030kg Transport : UN number : 2948 Chem class : 1 Pack group : II


Quantity 100mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10667242

  • £24.90 / 100mL


Estimated Shipment
15-03-2018
Add to basket


Specifications

Chemical Identifiers

3-Aminobenzotrifluoride
98-16-8
98%
C7H6F3N
161.13
3-(Trifluoromethyl)aniline
98%
98
98%
H2NC6H4CF3
MFCD00007797

Specifications

3-Aminobenzotrifluoride
98-16-8
Authentic
Glass bottle
1.3030g/mL
98%
5.0° to 6.0°C
C7H6F3N
H2NC6H4CF3
100mL
MFCD00007797
3-(Trifluoromethyl)aniline
98%
98
This product may darken on storage
1.4790 to 1.4810
Liquid
98%
Clear light yellow
187.0° to 193.0°C
161.13
85°C
12, 870
Videos
SDS