missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzophenone, 98%, Thermo Scientific™

Catalog Number p-3593277
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2835-78-1
C13H11NO
197.24
MFCD00017103
FUADXEJBHCKVBN-UHFFFAOYSA-N
3-aminobenzophenone, 3-benzoylaniline, methanone, 3-aminophenyl phenyl, 3-aminophenyl phenyl methanone, benzophenone, 3-amino, m-aminobenzophenone, 3-aminophenyl phenylmethanone, 3-aminophenyl phenyl ketone, 3-amino-benzophenon, 3-amino-benzophenone
17817
(3-aminophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)N
This item is not returnable. View return policy
2835-78-1
C13H11NO
197.24
MFCD00017103
FUADXEJBHCKVBN-UHFFFAOYSA-N
3-aminobenzophenone, 3-benzoylaniline, methanone, 3-aminophenyl phenyl, 3-aminophenyl phenyl methanone, benzophenone, 3-amino, m-aminobenzophenone, 3-aminophenyl phenylmethanone, 3-aminophenyl phenyl ketone, 3-amino-benzophenon, 3-amino-benzophenone
17817
(3-aminophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)N
CAS Min % 97.5
CAS Max % 100.0
Melting Point 84.0°C to 87.0°C
Color Yellow to Green
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C(O)C6H4NH2
Quantity 1 g
Formula Weight 197.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Aminobenzophenone
Show More Show Less
compliance-icons
Product Identifier
  • 3-Aminobenzophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'