missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Amino-6-bromopyrazine-2-carboxamide, 96%

Brand:  Alfa Aesar™ H63202.03

Code : NEW

Additional Details : CAS Number : 17890-77-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449768

  • £108.12 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

3-Amino-6-bromopyrazine-2-carboxamide
17890-77-6
217.02
3-Amino-6-bromopyrazine-2-carboxylic acid amide

Specifications

3-Amino-6-bromopyrazine-2-carboxamide
Solid
217.02
3-Amino-6-bromopyrazine-2-carboxylic acid amide
17890-77-6
96%
1g
Videos
SDS