missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-6-bromo-1H-indazole, 95%, Thermo Scientific Chemicals

Catalog Number 10883571
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
404827-77-6
C7H6BrN3
212.05
MFCD05665872
WLDHNAMVDBASAW-UHFFFAOYSA-N
3-amino-6-bromo-1h-indazole, 6-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 6-bromo, 6-bromo-1h-indazole-3-amine, pubchem17937, acmc-209jef, 6-bromo-3-amino-1h-indazole, 1h-indazol-3-amine,6-bromo, 3-amino-6-bromoindazole, 6-bromo-1h-indazole-3-ylamine
2786631
6-bromo-1H-indazol-3-amine
C1=CC2=C(C=C1Br)NN=C2N
This item is not returnable. View return policy
404827-77-6
C7H6BrN3
212.05
MFCD05665872
WLDHNAMVDBASAW-UHFFFAOYSA-N
3-amino-6-bromo-1h-indazole, 6-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 6-bromo, 6-bromo-1h-indazole-3-amine, pubchem17937, acmc-209jef, 6-bromo-3-amino-1h-indazole, 1h-indazol-3-amine,6-bromo, 3-amino-6-bromoindazole, 6-bromo-1h-indazole-3-ylamine
2786631
6-bromo-1H-indazol-3-amine
C1=CC2=C(C=C1Br)NN=C2N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 175°C to 178°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 212.05
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-6-bromo-1H-indazole
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-6-bromo-1H-indazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'