missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-tert-butyl-1H-pyrazole, 97%, ACROS Organics™

Catalog Number p-4812906
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
82560-12-1
C7H13N3
139.20
MFCD00067987,MFCD00082631
ZHBXGHWSVIBUCQ-UHFFFAOYSA-N
3-amino-5-tert-butylpyrazole, 3-tert-butyl-1h-pyrazol-5-amine, 5-amino-3-tert-butylpyrazole, 3-amino-5-tert-butyl-1h-pyrazole, 5-tert-butyl-2h-pyrazol-3-ylamine, 5-amino-3-t-butylpyrazole, 1h-pyrazol-3-amine, 5-1,1-dimethylethyl, 5-tert-butyl-2h-pyrazol-3-amine, 5-amino-3-tert-butyl-1h-pyrazole
522787
5-tert-butyl-1H-pyrazol-3-amine
CC(C)(C)C1=CC(N)=NN1
This item is not returnable. View return policy
82560-12-1
C7H13N3
139.20
MFCD00067987,MFCD00082631
ZHBXGHWSVIBUCQ-UHFFFAOYSA-N
3-amino-5-tert-butylpyrazole, 3-tert-butyl-1h-pyrazol-5-amine, 5-amino-3-tert-butylpyrazole, 3-amino-5-tert-butyl-1h-pyrazole, 5-tert-butyl-2h-pyrazol-3-ylamine, 5-amino-3-t-butylpyrazole, 1h-pyrazol-3-amine, 5-1,1-dimethylethyl, 5-tert-butyl-2h-pyrazol-3-amine, 5-amino-3-tert-butyl-1h-pyrazole
522787
5-tert-butyl-1H-pyrazol-3-amine
CC(C)(C)C1=CC(N)=NN1
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 139.2
Chemical Name or Material 3-Amino-5-tert-butyl-1H-pyrazole
compliance-icons
Product Identifier
  • 3-Amino-5-tert-butyl-1H-pyrazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'