missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-phenylpyrazole, 98%, Thermo Scientific™

Catalog Number p-2690289
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1572-10-7
C9H9N3
159.19
MFCD00191749
PWSZRRFDVPMZGM-UHFFFAOYSA-N
3-phenyl-1h-pyrazol-5-amine, 3-amino-5-phenylpyrazole, 5-amino-3-phenylpyrazole, 1h-pyrazol-3-amine, 5-phenyl, 5-phenyl-2h-pyrazol-3-amine, pyrazole, 5-amino-3-phenyl, 3-amino-5-phenyl-1h-pyrazole, 5-phenyl-2h-pyrazol-3-ylamine, 1h-pyrazol-5-amine, 3-phenyl, 5-phenyl-1h-pyrazol-3-ylamine
136655
5-phenyl-1H-pyrazol-3-amine
C1=CC=C(C=C1)C2=CC(=NN2)N
This item is not returnable. View return policy
1572-10-7
C9H9N3
159.19
MFCD00191749
PWSZRRFDVPMZGM-UHFFFAOYSA-N
3-phenyl-1h-pyrazol-5-amine, 3-amino-5-phenylpyrazole, 5-amino-3-phenylpyrazole, 1h-pyrazol-3-amine, 5-phenyl, 5-phenyl-2h-pyrazol-3-amine, pyrazole, 5-amino-3-phenyl, 3-amino-5-phenyl-1h-pyrazole, 5-phenyl-2h-pyrazol-3-ylamine, 1h-pyrazol-5-amine, 3-phenyl, 5-phenyl-1h-pyrazol-3-ylamine
136655
5-phenyl-1H-pyrazol-3-amine
C1=CC=C(C=C1)C2=CC(=NN2)N
CAS Min % 97.5
CAS Max % 100.0
Melting Point 118°C to 121°C
Color Brown to Red
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 24, 148
Formula Weight 159.19
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 3-Amino-5-phenylpyrazole
Show More Show Less
compliance-icons
Product Identifier
  • 5-phenyl-1H-pyrazol-3-amine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'