missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-methylpyrazole, 97%, ACROS Organics™

Brand:  Acros Organics 368031000

Code : 29

Additional Details : CAS Number : 31230-17-8 Weight : 0.15000kg


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10582062

  • £147.60 / 100g


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

3-Amino-5-methylpyrazole
31230-17-8
100.0
97%
97.12
5-Methyl-3-pyrazolamine
97%
96.0
97%
C4H7N3
MFCD00075180

Specifications

3-Amino-5-methylpyrazole
31230-17-8
96.0
Glass bottle
Light yellow to orange
97%
97%
C4H7N3
MFCD00075180
97%
Authentic
100.0
41.0° to 43.0°C
Solid
100g
>110°C
97.12
5-Methyl-3-pyrazolamine
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Amino-5-methylpyrazole

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS