missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-5-methyl-1H-pyrazole, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
3-Amino-5-methyl-1H-pyrazole was employed as beta-sheet template to investigate its interaction with ferrocenoyl-dipeptides. It is also used in the synthesis of 4-aryl-3,7,7-trimethyl-2,4,6,7,8,9-hexahydropyrazolo[3,4-b]quinolin-5-ones. It can react with 4-Ethoxymethylene-2-phenyl-4H-oxazol-5-one to get N-(2-Methyl-7-oxo-4,7-dihydro-pyrazolo[1,5-a]pyrimidin-6-yl)-benzamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Amino-5-methyl-1H-pyrazole was employed as beta-sheet template to investigate its interaction with ferrocenoyl-dipeptides. It is also used in the synthesis of 4-aryl-3,7,7-trimethyl-2,4,6,7,8,9-hexahydropyrazolo[3,4-b]quinolin-5-ones. It can react with 4-Ethoxymethylene-2-phenyl-4H-oxazol-5-one to get N-(2-Methyl-7-oxo-4,7-dihydro-pyrazolo[1,5-a]pyrimidin-6-yl)-benzamide.
Solubility
Soluble in Dichloromethane and Methanol.
Notes
Air Sensitive. Store at -20°C. Store under dry inert gas away from oxidizing agents and air.
3-Amino-5-methyl-1H-pyrazole was employed as beta-sheet template to investigate its interaction with ferrocenoyl-dipeptides. It is also used in the synthesis of 4-aryl-3,7,7-trimethyl-2,4,6,7,8,9-hexahydropyrazolo[3,4-b]quinolin-5-ones. It can react with 4-Ethoxymethylene-2-phenyl-4H-oxazol-5-one to get N-(2-Methyl-7-oxo-4,7-dihydro-pyrazolo[1,5-a]pyrimidin-6-yl)-benzamide.
Solubility
Soluble in Dichloromethane and Methanol.
Notes
Air Sensitive. Store at -20°C. Store under dry inert gas away from oxidizing agents and air.
Specifications
Melting Point | 42°C to 47°C |
Boiling Point | 212°C to 213°C (14 mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 5 g |
Beilstein | 1904 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in Dichloromethane and Methanol. |
Formula Weight | 97.12 |
Percent Purity | 97% |
Chemical Name or Material | 3-Amino-5-methyl-1H-pyrazole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'