missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-iodo-1H-indazole, 95%, Thermo Scientific™

Catalog Number p-4228315
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
88805-76-9
C7H6IN3
259.05
MFCD09261138
GBUVSJWTWOWRSL-UHFFFAOYSA-N
3-amino-5-iodo-1h-indazole, 1h-indazol-3-amine,5-iodo, 1h-indazol-3-amine, 5-iodo, 3-amino-5-iodoindazole, acmc-20f1u4, 5-iodo-1h-indazole-3-ylamine
20135868
5-iodo-1H-indazol-3-amine
NC1=NNC2=CC=C(I)C=C12
This item is not returnable. View return policy
88805-76-9
C7H6IN3
259.05
MFCD09261138
GBUVSJWTWOWRSL-UHFFFAOYSA-N
3-amino-5-iodo-1h-indazole, 1h-indazol-3-amine,5-iodo, 1h-indazol-3-amine, 5-iodo, 3-amino-5-iodoindazole, acmc-20f1u4, 5-iodo-1h-indazole-3-ylamine
20135868
5-iodo-1H-indazol-3-amine
NC1=NNC2=CC=C(I)C=C12
CAS Min % 94.0
CAS Max % 100.0
Melting Point 182°C to 188°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 259.05
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5-iodo-1H-indazole
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-5-iodo-1H-indazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'