missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-5-fluorobenzoic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Quantity | 250 mg |
Synonym | benzoic acid, 3-amino-5-fluoro-9ci, 3-amino-5-fluoro-benzoic acid, 5-amino-3-fluorobenzoic acid, 3-fluoro-5-aminobenzoic acid, benzoic acid,3-amino-5-fluoro, benzoic acid, 3-amino-5-fluoro |
Formula Weight | 155.13 |
Percent Purity | 95% |
Physical Form | Solid |
Chemical Name or Material | 3-Amino-5-fluorobenzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'