missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-5-(2-furyl)-1H-pyrazole, 97%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
Description
3-Amino-5-(2-furyl)-1H-pyrazole is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Amino-5-(2-furyl)-1H-pyrazole is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
3-Amino-5-(2-furyl)-1H-pyrazole is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Spécification
Melting Point | 124°C to 128°C |
Quantity | 250 mg |
Beilstein | 4399373 |
Solubility Information | Insoluble in water. |
Formula Weight | 149.15 |
Percent Purity | 97% |
Chemical Name or Material | 3-Amino-5-(2-furyl)-1H-pyrazole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'