missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methylbenzeneboronic acid, 98%, Thermo Scientific™

Catalog Number p-7047135
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
22237-12-3
C7H11BClNO2
187.43
MFCD01632201
BKFDOPYYSBQWIK-UHFFFAOYSA-N
3-amino-4-methylphenyl boronic acid, 3-amino-4-methylbenzeneboronic acid, 3-amino-4-methyl-phenyl boronic acid, 3-amino-p-tolylboronic acid, boronic acid, 3-amino-4-methylphenyl, acmc-20agxu, pubchem1745, 3-amino-4-methylphenylboranic acid, 3-amino4methyl phenyl boronic acid
2737803
(3-amino-4-methylphenyl)boronic acid
Cl.CC1=CC=C(C=C1N)B(O)O
This item is not returnable. View return policy
22237-12-3
C7H11BClNO2
187.43
MFCD01632201
BKFDOPYYSBQWIK-UHFFFAOYSA-N
3-amino-4-methylphenyl boronic acid, 3-amino-4-methylbenzeneboronic acid, 3-amino-4-methyl-phenyl boronic acid, 3-amino-p-tolylboronic acid, boronic acid, 3-amino-4-methylphenyl, acmc-20agxu, pubchem1745, 3-amino-4-methylphenylboranic acid, 3-amino4methyl phenyl boronic acid
2737803
(3-amino-4-methylphenyl)boronic acid
Cl.CC1=CC=C(C=C1N)B(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point ∼250°C
Quantity 1 g
Formula Weight 150.98
Percent Purity 98%
Chemical Name or Material 3-Amino-4-methylbenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'