missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methoxybenzenesulfonic acid, 98%, ACROS Organics™

Brand:  Acros Organics 342610050

Code : 29

Additional Details : CAS Number : 98-42-0 Weight : 0.06000kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10173341

  • £36.80 / 5g


Estimated Shipment
21-06-2018
Add to basket


Specifications

Chemical Identifiers

3-Amino-4-methoxybenzenesulfonic acid
98-42-0
100.0
98%
H3COC6H3(NH2)SO2OH
MFCD00035759
98%
97.5
≥97.5%
C7H9NO4S
203.21

Specifications

3-Amino-4-methoxybenzenesulfonic acid
98-42-0
100.0
Glass bottle
≥97.5%
C7H9NO4S
203.21
Off-white
98%
97.5
Authentic
Crystalline Powder
98%
H3COC6H3(NH2)SO2OH
MFCD00035759
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Amino-4-methoxybenzenesulfonic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS