missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Amino-4-methoxybenzenesulfonic acid, 98%

Brand:  Alfa Aesar™ H59256.14

Code : NEW

Additional Details : CAS Number : 98-42-0 Weight : 0.02500kg Transport : UN number : 2585 Chem class : 1a Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 15455508

  • £137.91 / 25g


Estimated Shipment date
from Supplier 20-03-2018
Add to basket

Specifications

Chemical Identifiers

3-Amino-4-methoxybenzenesulfonic acid
H2NC6H3(OCH3)SO3H
MFCD00035759
4-Methoxymetanilic acid; o-Anisidine-4-sulfonic acid
98-42-0
203.22
UN2585

Specifications

3-Amino-4-methoxybenzenesulfonic acid
Solid
305°C
203.22
White
25g
98-42-0
98%
H2NC6H3(OCH3)SO3H
MFCD00035759
UN2585
4-Methoxymetanilic acid; o-Anisidine-4-sulfonic acid
Videos
SDS