missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-amino-4-hydroxy-5-nitrobenzene-1-sulfonic acid hydrate, Thermo Scientific™
Click to view available options
Quantity:
5 g
10 g
25 g
Specifications
Assay Percent Range | 97% |
Quantity | 10 g |
Formula Weight | 252.21 |
Percent Purity | 97% |
Chemical Name or Material | 3-Amino-4-hydroxy-5-nitrobenzene-1-sulfonic acid hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'