missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Amino-4-fluorophenol, 98%

Brand:  Alfa Aesar™ H61829.03

Code : NEW

Additional Details : CAS Number : 62257-16-3 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15408418

  • £177.02 / 1g


Estimated Shipment date
from Supplier 23-07-2018
Add to basket

Specifications

Chemical Identifiers

3-Amino-4-fluorophenol
127.12
UN2811
62257-16-3
MFCD09033850

Specifications

3-Amino-4-fluorophenol
Powder
127.12
Yellow-brown
1g
62257-16-3
98%
MFCD09033850
UN2811
Videos
SDS