missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Amino-4-ethyl-1H-pyrazole, 98%

Brand:  Alfa Aesar™ H63199.MD

Code : NEW

Additional Details : CAS Number : 43024-15-3 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15439768

  • £27.10 / 250mg


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Description

Description

Applications
3-Amino-4-ethyl-1H-pyrazole is employed as a intermediate for chemical research and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

3-Amino-4-ethyl-1H-pyrazole
43024-15-3
111.15
MFCD06797570

Specifications

3-Amino-4-ethyl-1H-pyrazole
Solid
250mg
MFCD06797570
43024-15-3
98%
111.15
Videos
SDS