missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-carbethoxypyrazole, 99%, ACROS Organics™

£28.57

Chemical Identifiers

CAS 6994-25-8
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00005238
InChI Key YPXGHKWOJXQLQU-UHFFFAOYSA-N
Synonym ethyl 3-amino-1h-pyrazole-4-carboxylate, ethyl 3-amino-4-pyrazolecarboxylate, 3-amino-4-carbethoxypyrazole, 5-amino-1h-pyrazole-4-carboxylic acid ethyl ester, 3-amino-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl 3-aminopyrazole-4-carboxylate, 3-amino-4-ethoxycarbonylpyrazole, unii-9gy5ha1gaa, 3-amino-4-carbethoxypyrazol, 1h-pyrazole-4-carboxylic acid, 3-amino-, ethyl ester
PubChem CID 81472
IUPAC Name ethyl 5-amino-1H-pyrazole-4-carboxylate
SMILES CCOC(=O)C1=C(NN=C1)N
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10050580
SDS View Documents
Acros Organics
103310050
5g
£28.57
5g
View alternative products
Save Up to 
»
In stock, ready to ship: 1 Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

6994-25-8
155.16
YPXGHKWOJXQLQU-UHFFFAOYSA-N
81472
CCOC(=O)C1=C(NN=C1)N
C6H9N3O2
MFCD00005238
ethyl 3-amino-1h-pyrazole-4-carboxylate, ethyl 3-amino-4-pyrazolecarboxylate, 3-amino-4-carbethoxypyrazole, 5-amino-1h-pyrazole-4-carboxylic acid ethyl ester, 3-amino-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl 3-aminopyrazole-4-carboxylate, 3-amino-4-ethoxycarbonylpyrazole, unii-9gy5ha1gaa, 3-amino-4-carbethoxypyrazol, 1h-pyrazole-4-carboxylic acid, 3-amino-, ethyl ester
ethyl 5-amino-1H-pyrazole-4-carboxylate

Specifications

3-Amino-4-carbethoxypyrazole
Glass bottle
103.0°C to 107.0°C
99%
98.5
98.5% min. (HPLC)
MFCD00005238
YPXGHKWOJXQLQU-UHFFFAOYSA-N
ethyl 5-amino-1H-pyrazole-4-carboxylate
81472
Crystalline Powder
Authentic
White to Yellow
5g
6994-25-8
100.0
C6H9N3O2
ethyl 3-amino-1h-pyrazole-4-carboxylate, ethyl 3-amino-4-pyrazolecarboxylate, 3-amino-4-carbethoxypyrazole, 5-amino-1h-pyrazole-4-carboxylic acid ethyl ester, 3-amino-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl 3-aminopyrazole-4-carboxylate, 3-amino-4-ethoxycarbonylpyrazole, unii-9gy5ha1gaa, 3-amino-4-carbethoxypyrazol, 1h-pyrazole-4-carboxylic acid, 3-amino-, ethyl ester
CCOC(=O)C1=C(NN=C1)N
155.16
155.16
99%
SDS