missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-4-[(1-benzyl-2-methoxy-2-oxoethyl)amino]-4-oxobutanoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
100 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 50 g |
IUPAC Name | 3-amino-3-[(1-methoxy-1-oxo-3-phenylpropan-2-yl)carbamoyl]propanoic acid |
Formula Weight | 294.31 |
Percent Purity | 97% |
Chemical Name or Material | 3-Amino-4-[(1-benzyl-2-methoxy-2-oxoethyl)amino]-4-oxobutanoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'