missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-3-phenyl-1-propanol, 94%, Thermo Scientific Chemicals

Catalog Number p-4510456
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
14593-04-5
C9H13NO
151.21
MFCD00801124,MFCD01311768,MFCD01311791
SEQXIQNPMQTBGN-UHFFFAOYNA-N
3-amino-3-phenyl-1-propanol, dl-beta-phenylalaninol, 3-amino-3-phenyl-propan-1-ol, 3-phenyl-dl-beta-alaninol, 1-propanol, 3-amino-3-phenyl, benzenepropanol, gamma-amino, 1-amino-3-hydroxyprop-1-yl benzene, dl--phenylalaninol, 3-amino-3-phenylpropanol, acmc-1c4c0
203500
3-amino-3-phenylpropan-1-ol
NC(CCO)C1=CC=CC=C1
This item is not returnable. View return policy
14593-04-5
C9H13NO
151.21
MFCD00801124,MFCD01311768,MFCD01311791
SEQXIQNPMQTBGN-UHFFFAOYNA-N
3-amino-3-phenyl-1-propanol, dl-beta-phenylalaninol, 3-amino-3-phenyl-propan-1-ol, 3-phenyl-dl-beta-alaninol, 1-propanol, 3-amino-3-phenyl, benzenepropanol, gamma-amino, 1-amino-3-hydroxyprop-1-yl benzene, dl--phenylalaninol, 3-amino-3-phenylpropanol, acmc-1c4c0
203500
3-amino-3-phenylpropan-1-ol
NC(CCO)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 70.0°C to 77.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Linear Formula C6H5CH(NH2)CH2CH2OH
Quantity 500 mg
Beilstein 13,IV,1919
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol, ethanol and chloroform
Formula Weight 151.21
Percent Purity 94%
Physical Form Needles
Chemical Name or Material 3-Amino-3-phenyl-1-propanol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-3-phenyl-1-propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'