missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-phenylindenone, 98%, Thermo Scientific™

Catalog Number p-7081313
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1947-47-3
C15H11NO
221.259
MFCD00051614
HLEKBTIHDXNOQP-UHFFFAOYSA-N
3-amino-2-phenyl-1h-inden-1-one, 3-amino-2-phenylindenone, maybridge1_003093, acmc-1c032, 3-amino-2-phenyl-inden-1-one, 2-phenyl-3-amino-1h-indene-1-one, 1h-inden-1-one,3-amino-2-phenyl, 3-amino-2-phenyl-1h-inden-1-one, tech.
291219
3-amino-2-phenylinden-1-one
C1=CC=C(C=C1)C2=C(C3=CC=CC=C3C2=O)N
This item is not returnable. View return policy
1947-47-3
C15H11NO
221.259
MFCD00051614
HLEKBTIHDXNOQP-UHFFFAOYSA-N
3-amino-2-phenyl-1h-inden-1-one, 3-amino-2-phenylindenone, maybridge1_003093, acmc-1c032, 3-amino-2-phenyl-inden-1-one, 2-phenyl-3-amino-1h-indene-1-one, 1h-inden-1-one,3-amino-2-phenyl, 3-amino-2-phenyl-1h-inden-1-one, tech.
291219
3-amino-2-phenylinden-1-one
C1=CC=C(C=C1)C2=C(C3=CC=CC=C3C2=O)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 268°C to 274°C
Quantity 5 g
Beilstein 1875348
Formula Weight 221.26
Percent Purity 98%
Chemical Name or Material 3-Amino-2-phenylindenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'