missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-methylbenzyl alcohol, 97%, Thermo Scientific™

Catalog Number 11965291
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
83647-42-1
C8H11NO
137.18
UVYZMJMDIMWDNJ-UHFFFAOYSA-N
3-amino-2-methylphenyl methanol, 3-amino-2-methylbenzyl alcohol, benzenemethanol, 3-amino-2-methyl, 3-amino-2-methylbenzylalcohol, 3-amino-2-methylbenzenemethanol, pubchem19790, acmc-20a9oa, 3-amino-2-methylphenyl metanol, 3-amino-2-methyl benzyl alcohol, 3-amino-2-methyl-phenyl methanol
577175
(3-amino-2-methylphenyl)methanol
CC1=C(C=CC=C1N)CO
This item is not returnable. View return policy
83647-42-1
C8H11NO
137.18
UVYZMJMDIMWDNJ-UHFFFAOYSA-N
3-amino-2-methylphenyl methanol, 3-amino-2-methylbenzyl alcohol, benzenemethanol, 3-amino-2-methyl, 3-amino-2-methylbenzylalcohol, 3-amino-2-methylbenzenemethanol, pubchem19790, acmc-20a9oa, 3-amino-2-methylphenyl metanol, 3-amino-2-methyl benzyl alcohol, 3-amino-2-methyl-phenyl methanol
577175
(3-amino-2-methylphenyl)methanol
CC1=C(C=CC=C1N)CO
CAS Min % 96.0
CAS Max % 100.0
Melting Point 108.9°C to 109.7°C
Color Orange-Purple to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 137.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Amino-2-methylbenzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-2-methylbenzyl alcohol, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'