missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Amino-2-fluoro-4-methylpyridine, 98%

Brand:  Alfa Aesar™ H64758.03

Code : NEW

Additional Details : CAS Number : 173435-32-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15462089

  • £41.80 / 1g


Estimated Shipment date
from Supplier 06-07-2018
Add to basket

Specifications

Chemical Identifiers

3-Amino-2-fluoro-4-methylpyridine
126.13
13755262
173435-32-0
MFCD04112556
3-Amino-2-fluoro-4-picoline

Specifications

3-Amino-2-fluoro-4-methylpyridine
Solid
63° to 64°C
MFCD04112556
1g
3-Amino-2-fluoro-4-picoline
173435-32-0
98%
126.13
Off-white
13755262
Videos
SDS