missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-chloropyridine 96%, ACROS Organics™

Brand:  Acros Organics 103420500

Code : 29

Additional Details : CAS Number : 6298-19-7 Weight : 0.10000kg


Quantity 50g
Packaging Glass bottle

 View more versions of this product

Product Code. 10626862

  • £51.20 / 50g


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

3-Amino-2-chloropyridine
6298-19-7
100.0
96%
Cl(C5H3N)NH2
MFCD00006238
96%
95.0
96%
C5H5ClN2
128.56

Specifications

3-Amino-2-chloropyridine
Glass bottle
Authentic
100.0
Off-white to yellow to pink
Crystalline Powder, Crystals, and/or Chunks
96%
Cl(C5H3N)NH2
MFCD00006238
96%
6298-19-7
95.0
77.0° to 82.0°C
50g
96%
C5H5ClN2
128.56
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Amino-2-chloropyridine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS