missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3-Amino-2-chloro-6-methylpyridine, 98%
Click to view available options
Quantity:
1 g
5 g
Specifications
Melting Point | 64°C to 72°C |
Color | White to Yellow |
Quantity | 5 g |
UN Number | UN2811 |
Beilstein | 114030 |
Formula Weight | 142.59 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 3-Amino-2-chloro-6-methylpyridine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'