missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-chloro-5-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog Number 15429276
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
72587-18-9
C6H4ClF3N2
196.557
MFCD07375383
HABKYPVMMHBOIW-UHFFFAOYSA-N
2-chloro-5-trifluoromethyl pyridin-3-amine, 2-chloro-5-trifluoromethyl-3-pyridinamine, 3-amino-2-chloro-5-trifluoromethyl pyridine, 3-amino-2-chloro-5-trifluoromethylpyridine, 2-chloro-3-amino-5-trifluoromethyl pyridine
12919757
2-chloro-5-(trifluoromethyl)pyridin-3-amine
C1=C(C=NC(=C1N)Cl)C(F)(F)F
This item is not returnable. View return policy
72587-18-9
C6H4ClF3N2
196.557
MFCD07375383
HABKYPVMMHBOIW-UHFFFAOYSA-N
2-chloro-5-trifluoromethyl pyridin-3-amine, 2-chloro-5-trifluoromethyl-3-pyridinamine, 3-amino-2-chloro-5-trifluoromethyl pyridine, 3-amino-2-chloro-5-trifluoromethylpyridine, 2-chloro-3-amino-5-trifluoromethyl pyridine
12919757
2-chloro-5-(trifluoromethyl)pyridin-3-amine
C1=C(C=NC(=C1N)Cl)C(F)(F)F

It's employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It’s employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color White to Brown
Quantity 250 mg
Beilstein 19757170
Solubility Information Slightly soluble in water.
Formula Weight 196.56
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Amino-2-chloro-5-(trifluoromethyl)pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'