missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Amino-2,6-dimethylpyridine, 97%

Brand:  Alfa Aesar™ L19842.06

Code : NEW

Additional Details : CAS Number : 3430-33-9 Weight : 0.00500kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 11309835

  • £97.70 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Description

Description

Applications
3-Amino-2,6-dimethylpyridine is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

3-Amino-2,6-dimethylpyridine
C7H10N2
MFCD01646409
2424
3430-33-9
122.17
UN2811
3-Amino-2,6-lutidine

Specifications

3-Amino-2,6-dimethylpyridine
121° to 123°C
97%
C7H10N2
MFCD01646409
2424
3430-33-9
230°C
5g
122.17
UN2811
3-Amino-2,6-lutidine
Videos
SDS