missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-2,4,6-triiodobenzoic acid, 99% (dry wt.), water <4%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
3-Amino-2,4,6-triiodobenzoic acid is used to prepare polymeric hydrogel based particles used in vascular embolization. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Amino-2,4,6-triiodobenzoic acid is used to prepare polymeric hydrogel based particles used in vascular embolization. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Insoluble in water.
Notes
Protect from exposure to light. Store in the dark. Incompatible with light and oxidizing agents. Store at room temperature.
3-Amino-2,4,6-triiodobenzoic acid is used to prepare polymeric hydrogel based particles used in vascular embolization. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Insoluble in water.
Notes
Protect from exposure to light. Store in the dark. Incompatible with light and oxidizing agents. Store at room temperature.
Specifications
Melting Point | ∼196°C (decomposition) |
Quantity | 5 g |
Sensitivity | Light sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 514.83 |
Percent Purity | 99% |
Assay | (dry wt.), may cont. up to 1 mole of water |
Chemical Name or Material | 3-Amino-2,4,6-triiodobenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'