missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1H-1,2,4-triazole, 95%, ACROS Organics™

Brand:  Acros Organics 264571000

Code : 29

Additional Details : CAS Number : 61-82-5 Weight : 0.15000kg Transport : UN number : 3077 Chem class : 9 Pack group : III

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10702681

  • £50.20 / 100g


EU Stock 3
Add to basket

Specifications

Chemical Identifiers

3-Amino-1H-1, 2, 4-triazole
61-82-5
100.0
95%
84.08
95%
94.0
≥94%
C2H4N4
MFCD00005230

Specifications

3-Amino-1H-1, 2, 4-triazole
61-82-5
100.0
Glass bottle
≥94%
C2H4N4
84.08
MFCD00005230
26, 137
95%
94.0
Authentic
Powder or Flakes
95%
White to off-white or yellow
147° to 156°C
100g
15, 485
Videos
SDS