missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-propanol, 99%, Thermo Scientific Chemicals

Catalog Number 10616204
Click to view available options
Quantity:
5 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
156-87-6
MFCD00008223
WUGQZFFCHPXWKQ-UHFFFAOYSA-N
3-aminopropanol, 3-amino-1-propanol, propanolamine, 1-propanol, 3-amino, 3-propanolamine, 1-amino-3-propanol, 3-hydroxypropylamine, 3-aminopropyl alcohol, 1,3-propanolamine, beta-alaninol
9086
3-aminopropan-1-ol
C(CN)CO
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
156-87-6
MFCD00008223
WUGQZFFCHPXWKQ-UHFFFAOYSA-N
3-aminopropanol, 3-amino-1-propanol, propanolamine, 1-propanol, 3-amino, 3-propanolamine, 1-amino-3-propanol, 3-hydroxypropylamine, 3-aminopropyl alcohol, 1,3-propanolamine, beta-alaninol
9086
3-aminopropan-1-ol
C(CN)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 10.0°C to 12.0°C
Color Colorless to Yellow
Density 0.9820g/mL
Boiling Point 187.0°C to 188.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C3H9NO
Linear Formula H2N(CH2)3OH
Refractive Index 1.4595 to 1.4615
Quantity 2.5 L
Beilstein 04, 288
Specific Gravity 0.982
Solubility Information Solubility in water: freely soluble. Other solubilities: miscible with alcohols,esters, chloroform, acetone, slightly soluble in arom. and aliph. hydrocarbons
Molecular Weight (g/mol) 75.11
Viscosity 36 mPa.s (20°C)
Formula Weight 75.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Amino-1-propanol, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Amino-1-propanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'