missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-propanesulfonic acid, 95%, Thermo Scientific Chemicals

Catalog Number p-3756825
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3687-18-1
C3H9NO3S
139.17
MFCD00008225
SNKZJIOFVMKAOJ-UHFFFAOYSA-N
homotaurine, 3-amino-1-propanesulfonic acid, tramiprosate, alzhemed, 3-aminopropanesulphonic acid, 3aps, 1-propanesulfonic acid, 3-amino, cerebril, 3-aminopropanesulfonic acid, 3-aminopropane sulfonic acid
1646
CHEBI:1457
3-aminopropane-1-sulfonic acid
NCCCS(O)(=O)=O
This item is not returnable. View return policy
3687-18-1
C3H9NO3S
139.17
MFCD00008225
SNKZJIOFVMKAOJ-UHFFFAOYSA-N
homotaurine, 3-amino-1-propanesulfonic acid, tramiprosate, alzhemed, 3-aminopropanesulphonic acid, 3aps, 1-propanesulfonic acid, 3-amino, cerebril, 3-aminopropanesulfonic acid, 3-aminopropane sulfonic acid
1646
CHEBI:1457
3-aminopropane-1-sulfonic acid
NCCCS(O)(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point >270°C
Color Blue to White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula H2N(CH2)3SO3H
Quantity 1 g
Merck Index 15, 9728
Formula Weight 139.17
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-1-propanesulfonic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'