missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-methyl-1H-pyrazole-4-carbaldehyde, 97%, Thermo Scientific™

Catalog Number p-3485442
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
886851-66-7
C5H7N3O
125.13
MFCD00460466
UGWUYEXHMDPAMR-UHFFFAOYSA-N
3-amino-1-methyl-1h-pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde,3-amino-1-methyl, 1h-pyrazole-4-carboxaldehyde, 3-amino-1-methyl, 3-amino-1-methyl-1h-pyrazole-4-carboxaldehyde
18525942
3-amino-1-methylpyrazole-4-carbaldehyde
CN1C=C(C=O)C(N)=N1
This item is not returnable. View return policy
886851-66-7
C5H7N3O
125.13
MFCD00460466
UGWUYEXHMDPAMR-UHFFFAOYSA-N
3-amino-1-methyl-1h-pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde,3-amino-1-methyl, 1h-pyrazole-4-carboxaldehyde, 3-amino-1-methyl, 3-amino-1-methyl-1h-pyrazole-4-carboxaldehyde
18525942
3-amino-1-methylpyrazole-4-carbaldehyde
CN1C=C(C=O)C(N)=N1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 125.13
Percent Purity 97%
Chemical Name or Material 3-Amino-1-methyl-1H-pyrazole-4-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'