missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-methyl-1H-indazole, 95%, Thermo Scientific Chemicals

Catalog Number p-4228321
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
60301-20-4
C8H9N3
147.18
MFCD09054755
NYLGITXFVVEBLZ-UHFFFAOYSA-N
1-methyl-1h-indazol-3-amine, 3-amino-1-methylindazole, 3-amino-1-methyl-1h-indazole, 1h-indazol-3-amine, 1-methyl, n-methyl-3-aminoindazole, 1-methyl-1h-3-indazoleamine, 1-methyl-1h-indazole-3-amine, 1-methyl-1h-indazol-3-ylamine, 1-methyl-1h-indazole-3-ylamine
12291317
CN1N=C(N)C2=CC=CC=C12
This item is not returnable. View return policy
60301-20-4
C8H9N3
147.18
MFCD09054755
NYLGITXFVVEBLZ-UHFFFAOYSA-N
1-methyl-1h-indazol-3-amine, 3-amino-1-methylindazole, 3-amino-1-methyl-1h-indazole, 1h-indazol-3-amine, 1-methyl, n-methyl-3-aminoindazole, 1-methyl-1h-3-indazoleamine, 1-methyl-1h-indazole-3-amine, 1-methyl-1h-indazol-3-ylamine, 1-methyl-1h-indazole-3-ylamine
12291317
CN1N=C(N)C2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 95°C to 99°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
IUPAC Name 1-methyl-1H-indazol-3-amine
Formula Weight 147.18
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-1-methyl-1H-indazole
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-1-methyl-1H-indazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'