missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1,2-propanediol, 98%, ACROS Organics™

Brand:  Acros Organics 104420250

Code : 29

Additional Details : CAS Number : 616-30-8 Weight : 0.07500kg Transport : UN number : 2735 Chem class : 8 Pack group : III


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10774591

  • £44.20 / 25g


EU Stock 11
Add to basket


Specifications

Chemical Identifiers

3-Amino-1, 2-propanediol
616-30-8
100.0
98%
H2NCH2CH(OH)CH2OH
MFCD00008140
98%
97.5
98%
C3H9NO2
91.11

Specifications

3-Amino-1, 2-propanediol
616-30-8
97.5
Glass bottle
Solubility in water: soluble in water. Other solubilities: soluble in alcohols
1.1700g/mL
264.0° to 265.0°C (739.0 mmHg)
25g
98%
C3H9NO2
91.11
98%
Authentic
100.0
1.4900 to 1.4940
0.5% max. (K.F.)
Clear colorless to pale yellow
Viscous Liquid
155°C
98%
H2NCH2CH(OH)CH2OH
MFCD00008140
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Amino-1,2-propanediol

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS