missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1,2-propanediol, 98%, Thermo Scientific Chemicals

Catalog Number 10666092
Click to view available options
Quantity:
5 g
25 g
100 g
250 g
Packaging:
Glass bottle
616-30-8
91.11
MFCD00008140
KQIGMPWTAHJUMN-UHFFFAOYSA-N
3-amino-1,2-propanediol, 2,3-dihydroxypropylamine, 1,2-propanediol, 3-amino, 1-amino-2,3-propanediol, 1-aminoglycerol, 2,3-propandiol-1-amine, 1-aminopropanediol, 3-aminopropan-1,2-diol, 3-amino-1,2-proopanediol, +/--3-amino-1,2-propanediol
73561
3-aminopropane-1,2-diol
C(C(CO)O)N
616-30-8
91.11
MFCD00008140
KQIGMPWTAHJUMN-UHFFFAOYSA-N
3-amino-1,2-propanediol, 2,3-dihydroxypropylamine, 1,2-propanediol, 3-amino, 1-amino-2,3-propanediol, 1-aminoglycerol, 2,3-propandiol-1-amine, 1-aminopropanediol, 3-aminopropan-1,2-diol, 3-amino-1,2-proopanediol, +/--3-amino-1,2-propanediol
73561
3-aminopropane-1,2-diol
C(C(CO)O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1700g/mL
Boiling Point 264.0°C to 265.0°C (739.0 mmHg)
Flash Point 155°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C3H9NO2
Linear Formula H2NCH2CH(OH)CH2OH
Refractive Index 1.4900 to 1.4940
Quantity 100 g
Beilstein 04,301
Specific Gravity 1.17
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohols
Viscosity 3 Pas (20°C)
Formula Weight 91.11
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material 3-Amino-1, 2-propanediol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-1,2-propanediol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'