missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Acetylpyridine p-toluenesulfonylhydrazone, 97%

Brand:  Alfa Aesar™ H65233.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15492489

  • £26.29 / 1g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

3-Acetylpyridine p-toluenesulfonylhydrazone
289.35
MFCD01230698

Specifications

3-Acetylpyridine p-toluenesulfonylhydrazone
97%
MFCD01230698
1g
Solid
289.35
White to pale yellow
Videos
SDS