missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylphenyl isocyanate, 97%, Thermo Scientific™

Catalog Number p-49623
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
23138-64-9
C9H7NO2
161.16
MFCD00013866
JASTZOAHAWBRLM-UHFFFAOYSA-N
1-3-isocyanatophenyl ethanone, 3-acetylphenyl isocyanate, 3-acetylphenylisocyanate, 3-isocyanatoacetophenone, ethanone,1-3-isocyanatophenyl, ethanone, 1-3-isocyanatophenyl, 3-acetylbenzenisocyanate, 3'-isocyanatoacetophenone, acmc-1cjm8, 3'-isocyanato-acetophenone
312718
1-(3-isocyanatophenyl)ethanone
CC(=O)C1=CC=CC(=C1)N=C=O
This item is not returnable. View return policy
23138-64-9
C9H7NO2
161.16
MFCD00013866
JASTZOAHAWBRLM-UHFFFAOYSA-N
1-3-isocyanatophenyl ethanone, 3-acetylphenyl isocyanate, 3-acetylphenylisocyanate, 3-isocyanatoacetophenone, ethanone,1-3-isocyanatophenyl, ethanone, 1-3-isocyanatophenyl, 3-acetylbenzenisocyanate, 3'-isocyanatoacetophenone, acmc-1cjm8, 3'-isocyanato-acetophenone
312718
1-(3-isocyanatophenyl)ethanone
CC(=O)C1=CC=CC(=C1)N=C=O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 33°C to 34°C
Color Yellow to Brown
Density 1.1700g/mL
Boiling Point 155°C (4.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3COC6H4NCO
Refractive Index 1.5610 to 1.563
Quantity 1 g
Specific Gravity 1.17
Formula Weight 161.16
Percent Purity 97%
Physical Form Liquid After Melting
Chemical Name or Material 3-Acetylphenyl isocyanate, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'