missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylindole, 97%, Thermo Scientific Chemicals

Catalog Number p-49615
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
703-80-0
C10H9NO
159.19
MFCD00005626
VUIMBZIZZFSQEE-UHFFFAOYSA-N
3-acetylindole, 1-1h-indol-3-yl ethanone, 3-acetyl-1h-indole, ethanone, 1-1h-indol-3-yl, acetyl-3-indole, 1-1h-indol-3-yl-ethanone, ketone, indol-3-yl methyl, 3-acetyl indole, indol-3-yl methyl ketone, 1-1h-indol-3-yl ethan-1-one
12802
1-(1H-indol-3-yl)ethanone
CC(=O)C1=CNC2=CC=CC=C21
This item is not returnable. View return policy
703-80-0
C10H9NO
159.19
MFCD00005626
VUIMBZIZZFSQEE-UHFFFAOYSA-N
3-acetylindole, 1-1h-indol-3-yl ethanone, 3-acetyl-1h-indole, ethanone, 1-1h-indol-3-yl, acetyl-3-indole, 1-1h-indol-3-yl-ethanone, ketone, indol-3-yl methyl, 3-acetyl indole, indol-3-yl methyl ketone, 1-1h-indol-3-yl ethan-1-one
12802
1-(1H-indol-3-yl)ethanone
CC(=O)C1=CNC2=CC=CC=C21

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 185°C to 191°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Beilstein 21, 316
Formula Weight 159.19
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Acetylindole, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'