missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Acetylbenzo[b]thiophene, 98%

Brand:  Alfa Aesar™ A10330.14

Code : NEW

Additional Details : CAS Number : 1128-05-8 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11444103

  • £445.09 / 25g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

3-Acetylbenzo[b]thiophene
C10H8OS
MFCD00051638
3-Acetylthianaphthene
1128-05-8
176.23
122587

Specifications

3-Acetylbenzo[b]thiophene
98%
176.23
122587
165° to 170°C (10mmHg)
25g
1128-05-8
C10H8OS
MFCD00051638
3-Acetylthianaphthene
61° to 66°C
Videos
SDS