missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Acetylbenzo[b]thiophene, 98%

Brand:  Alfa Aesar™ A10330.06

Code : NEW

Additional Details : CAS Number : 1128-05-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11434103

  • £110.77 / 5g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Acetylbenzo[b]thiophene
C10H8OS
MFCD00051638
3-Acetylthianaphthene
1128-05-8
176.23
122587

Specifications

3-Acetylbenzo[b]thiophene
98%
176.23
122587
165° to 170°C (10mmHg)
5g
1128-05-8
C10H8OS
MFCD00051638
3-Acetylthianaphthene
61° to 66°C
Videos
SDS