missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-2,4-dimethylpyrrole, 98%, Thermo Scientific Chemicals

Catalog Number p-4509359
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2386-25-6
C8H11NO
137.18
VGZCKCJMYREIKA-UHFFFAOYSA-N
3-acetyl-2,4-dimethylpyrrole, 2,4-dimethyl-3-acetylpyrrole, 1-2,4-dimethyl-1h-pyrrol-3-yl ethanone, 2,4-dimethylpyrrol-3-yl methyl ketone, ethanone, 1-2,4-dimethyl-1h-pyrrol-3-yl, ketone, 2,4-dimethylpyrrol-3-yl methyl, 1-2,4-dimethyl-1h-pyrrol-3-yl ethan-1-one, 2,4-dimethyl-3-acetyl-pyrrole, 3-acetyl-2,5-dimethyl-pyrrole, 1-2,4-dimethyl-1h-pyrrol-3-yl-ethanone
15163
1-(2,4-dimethyl-1H-pyrrol-3-yl)ethanone
CC1=CNC(=C1C(=O)C)C
This item is not returnable. View return policy
2386-25-6
C8H11NO
137.18
VGZCKCJMYREIKA-UHFFFAOYSA-N
3-acetyl-2,4-dimethylpyrrole, 2,4-dimethyl-3-acetylpyrrole, 1-2,4-dimethyl-1h-pyrrol-3-yl ethanone, 2,4-dimethylpyrrol-3-yl methyl ketone, ethanone, 1-2,4-dimethyl-1h-pyrrol-3-yl, ketone, 2,4-dimethylpyrrol-3-yl methyl, 1-2,4-dimethyl-1h-pyrrol-3-yl ethan-1-one, 2,4-dimethyl-3-acetyl-pyrrole, 3-acetyl-2,5-dimethyl-pyrrole, 1-2,4-dimethyl-1h-pyrrol-3-yl-ethanone
15163
1-(2,4-dimethyl-1H-pyrrol-3-yl)ethanone
CC1=CNC(=C1C(=O)C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 136.0°C to 139.0°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 137.18
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 3-Acetyl-2, 4-dimethylpyrrole
compliance-icons
Product Identifier
  • 3-Acetyl-2,4-dimethylpyrrole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'