missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Acetyl-1-(phenylsulfonyl)pyrrole, 98%

Brand:  Alfa Aesar™ H55751.14

Code : NEW

Additional Details : CAS Number : 81453-98-7 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15492088

  • £359.45 / 25g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Acetyl-1-(phenylsulfonyl)pyrrole
81453-98-7
249.28
MFCD00067752

Specifications

3-Acetyl-1-(phenylsulfonyl)pyrrole
Solid
249.28
96° to 99°C
81453-98-7
98%
MFCD00067752
25g
Videos
SDS