missing translation for 'onlineSavingsMsg'
Learn More

3-Acetamidophenol, 99+%, Thermo Scientific Chemicals

Catalog Number p-47670
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
621-42-1
C8H9NO2
151.16
MFCD00002263
QLNWXBAGRTUKKI-UHFFFAOYSA-N
3-acetamidophenol, n-3-hydroxyphenyl acetamide, metacetamol, 3'-hydroxyacetanilide, m-acetamidophenol, 3-hydroxyacetanilide, n-acetyl-m-aminophenol, m-hydroxyacetanilide, 3-acetylamino phenol, metalid
12124
CHEBI:76987
N-(3-hydroxyphenyl)acetamide
CC(=O)NC1=CC(=CC=C1)O
This item is not returnable. View return policy
621-42-1
C8H9NO2
151.16
MFCD00002263
QLNWXBAGRTUKKI-UHFFFAOYSA-N
3-acetamidophenol, n-3-hydroxyphenyl acetamide, metacetamol, 3'-hydroxyacetanilide, m-acetamidophenol, 3-hydroxyacetanilide, n-acetyl-m-aminophenol, m-hydroxyacetanilide, 3-acetylamino phenol, metalid
12124
CHEBI:76987
N-(3-hydroxyphenyl)acetamide
CC(=O)NC1=CC(=CC=C1)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 146°C to 149°C
Color Gray or Tan to White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3CONHC6H4OH
Quantity 25 g
Beilstein 13, 415
Solubility Information Solubility in water: freely soluble in hot water
Formula Weight 151.16
Percent Purity 99+%
Physical Form Crystalline Powder, Crystals or Needles
Chemical Name or Material 3-Acetamidophenol, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Acetamidophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'